Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-STRN3 Mouse Monoclonal Antibody [clone: S68]
Catalog # ABNOMAB2291
Supplier:  Abnova
Anti-STRN3 Mouse Monoclonal Antibody [clone: S68]
Catalog # ABNOMAB2291
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Striatin, Calmodulin Binding Protein 3
  • Clonality:
    Monoclonal
  • Clone:
    S68
  • Conjugation:
    Unconjugated
  • Host:
    Mouse
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1
  • Reactivity:
    Human,
    Rat,
    Mouse
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Form:
    Ascites
  • Gene ID:
    29966
  • Antigen synonyms:
    STRN3 |Cell-cycle autoantigen SG2NA |SG2NA |S/G2 antigen |GS2NA
  • Amino acid number:
    200 to 300
  • Storage buffer:
    0,02% sodium azide
  • Storage temperature:
    Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to amino acids 200-300 of human STRN3.
  • Size:
    100 μl
  • Pk:
    100 µl

Specifications

About this item

Mouse monoclonal antibody raised against synthetic peptide of STRN3.

Recommended Dilutions: Immunofluorescence: 1:5000; Western Blot: 1:250-1:2000

Type: Primary
Antigen: STRN3
Clonality: Monoclonal
Clone: S68
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat