Anti-DHX58 Rabbit Polyclonal Antibody
Catalog # ABNOPAB13437
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:DEXH (Asp-Glu-X-His) box polypeptide 58
- Antigen symbol:DHX58
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:79132
- Antigen synonyms:RLR-3|D11LGP2|D11lgp2e|Probable ATP-dependent RNA helicase DHX58|LGP2|D11LGP2E|Probable ATP-dependent helicase LGP2
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to N-terminus 14 amino acids of human DHX58.
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of DHX58.
Recommended Dilutions: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: DHX58
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human, Mouse, Rat