Order Entry
United Kingdom
Orders LinkContactUsLinkComponent
Anti-ZFX Rabbit Polyclonal Antibody

Anti-ZFX Rabbit Polyclonal Antibody

100 μl
Catalog # ABNOPAB20245
Supplier:   Abnova
Anti-ZFX Rabbit Polyclonal Antibody
Catalog # ABNOPAB20245
Supplier:  Abnova
Anti-ZFX Rabbit Polyclonal Antibody
Anti-ZFX Rabbit Polyclonal Antibody

100 μl

Price
£578.00
Log in for availability
1 * 100 µl
Product Details & Documents

Rabbit polyclonal antibody raised against recombinant ZFX.

  • Recommended Dilutions: Immunohistochemistry: 1:200-1:500; Immunofluorescence: 1-4 μg/ml

Type: Primary
Antigen: ZFX
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human

Documents

Specifications
  • Size:
    100 μl
  • Western blot:
    Yes
  • Clonality:
    Polyclonal
  • Purification:
    Antigen affinity purification
  • Antibody type:
    Primary
  • ImmunoFluorescence:
    Yes
  • Isotype:
    IgG
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Host:
    Rabbit
  • Sequence:
    VSLAHCTVPDDVLASDITSASMSMPEHVLTGDSIHVSDVGHVGHVGHVEHVVHDSVVEAEIVTDPLTTDVVSEEVL
  • Cross adsorption:
    No
  • Antigen name:
    Zinc Finger Protein, X-linked
  • Cat. No.:
    ABNOPAB20245
  • Reactivity:
    Human
  • Format:
    Liquid
  • Gene ID:
    7543
  • Antigen synonyms:
    Zinc finger X-chromosomal protein|ZNF926|ZFX
  • Immunogen:
    Recombinant protein corresponding to amino acids of human ZFX.
  • ImmunoChemistry:
    Yes
  • Conjugation:
    Unconjugated
  • Antigen symbol:
    ZFX
  • Storage buffer:
    PBS, pH 7,5 (40% glycerol, 0,02% sodium azide)
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.