Anti-S100 A12 Rabbit Polyclonal Antibody
Catalog # ABNOPAB12047
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:S100 Calcium Binding Protein A12
- Antigen symbol:S100A12
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Form:Antiserum
- Gene ID:6283
- Antigen synonyms:ENRAGE|CAGC|CGRP|MRP-6|p6|CAAF1|MRP6
- Storage buffer:0,1% sodium azide
- Storage temperature:Store at 4 °C for short term. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human S100A12.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against full length recombinant S100A12.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: S100 A12
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human