Anti-TNNT2 Mouse Monoclonal Antibody [clone: 7E7]
Catalog # ABNOMAB3100
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Troponin T Type 2
- Antigen symbol:TNNT2
- Clonality:Monoclonal
- Clone:7E7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG
- Reactivity:Pig,Monkey,Human,Bovine,Dog,Rat,Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:7139
- Antigen synonyms:TnTC|CMPD2|LVNC6|CMH2|cTnT|CMD1D|RCM3
- Storage buffer:PBS, pH 7,4 (0,1% sodium azide)
- Storage temperature:Store at 4 °C.
- Immunogen:Human TNNT2 complex.
- Size:200 μg
- Pk:200 µg
Specifications
About this item
Mouse monoclonal antibody raised against TNNT2.
Type: Primary
Antigen: TNNT2
Clonality: Monoclonal
Clone: 7E7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Bovine, Dog, Human, Monkey, Mouse, Pig, Rat