Anti-HCRTR1 Rabbit Polyclonal Antibody
Catalog # ABNOPAB14322
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:hypocretin (orexin) receptor 1
- Antigen symbol:HCRTR1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Bovine,Rat
- Environmentally Preferable:
- Cross adsorption:No
- Format:Lyophilized
- Gene ID:282248
- Amino acid number:12 to 33
- Storage buffer:PBS
- Sequence:RLYELLHGAGNHAAGILTL
- Storage temperature:Store at 4 °C on dry atmosphere. After reconstitution with deionized water, store at –20 °C or lower. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide (conjugated with KLH) corresponding to amino acids 12-33 of bovine HCRTR1.
- Size:100 μl
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of HCRTR1.
Recommended Dilutions: Immunohistochemistry: 1:1000-1:2000
Type: Primary
Antigen: HCRTR1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Bovine, Rat