Anti-LIME Mouse Monoclonal Antibody [clone: mLIME-05]
ABNOMAB8509
: Abnova
- Antibody type:Primary
- Antigen name:Lck interacting transmembrane adaptor 1
- Antigen symbol:LIME
- Clonality:Monoclonal
- Clone:mLIME-05
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG2a
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:72699
- Antigen synonyms:lck-interacting membrane protein|dJ583P15.4|LIME|LP8067|lck-interacting molecule|RP4-583P15.5|FLJ20406|lck-interacting transmembrane adapter 1
- Amino acid number:86 to 202
- Storage buffer:PBS, pH 7,4 (15 mM sodium azide)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 86-202 of mouse Lime1.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against partial recombiant Lime1.
Recommended Dilutions: Western Blot: 1-5 ?g/ml
Type: Primary
Antigen: LIME
Clonality: Monoclonal
Clone: mLIME-05
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Mouse