Anti-Oval Cell Marker Rat Monoclonal Antibody [clone: OC2-3C7]
ABNOMAB7741
: Abnova
- Antibody type:Primary
- Antigen name:Oval Cell Marker
- Clonality:Monoclonal
- Clone:OC2-3C7
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:Mouse
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Form:Tissue Culture Supernatant
- Storage buffer:0,09% sodium azide
- Storage temperature:Store at 4 °C. Do not freeze.
- Immunogen:Native purified oval cell marker from nonparenchymal cells from DDC-treated mice to induce the oval cell response.
- Size:100 μl
- Pk:100 µg
Rat monoclonal antibody raised against native oval cell marker.
Recommended Dilutions: Flow Cytometry: 1:50-1:100; Immunohistochemistry (Frozen sections): 1:100
Type: Primary
Antigen: Oval Cell Marker
Clonality: Monoclonal
Clone: OC2-3C7
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG1
Reactivity: Mouse