Anti-VSIG1 Mouse Monoclonal Antibody (PE (Phycoerythrin)) [clone: CT1]
ABNOMAB5858
: Abnova
- Antibody type:Primary
- Antigen name:V-set and immunoglobulin domain containing 1
- Antigen symbol:VSIG1
- Clonality:Monoclonal
- Clone:CT1
- Conjugation:PE (Phycoerythrin)
- Flow cytometry:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG1 kappa
- Reactivity:Chicken
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:414795
- Antigen synonyms:CHT1
- Storage buffer:PBS (0,1 mg/mL sodium azide)
- Storage temperature:Store in the dark at 4 °C. Do not freeze. Avoid prolonged exposure to light. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Native purified from chicken thymocytes.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against native VSIG1.
Recommended Dilutions: Flow Cytometry: 0.2 ?g/10^6 cells
Type: Primary
Antigen: VSIG1
Clonality: PE (Phycoerythrin)
Clone: Monoclonal
Conjugation: CT1
Epitope:
Host: IgG1 kappa
Isotype: Mouse
Reactivity: Chicken