Anti-ORAI3 Mouse Monoclonal Antibody [clone: 1B4F1]
ABNOMAB8085
: Abnova
- Antibody type:Primary
- Antigen name:ORAI calcium release-activated calcium modulator 3
- Antigen symbol:ORAI3
- Clonality:Monoclonal
- Clone:1B4F1
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:
- Reactivity:Human,Rat
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:93129
- Antigen synonyms:TMEM142C|Protein orai-3|Transmembrane protein 142C|ORAI3
- Storage buffer:PBS (0,02% sodium azide)
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to C-terminus 19 amino acids of human ORAI3.
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against synthetic peptide of ORAI3.
Recommended Dilutions: Western Blot: 2-4 ?g/ml
Type: Primary
Antigen: ORAI3
Clonality: Monoclonal
Clone: 1B4F1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human, Rat