Anti-NELFB Mouse Monoclonal Antibody
ABNOMAB8174
: Abnova
- Antibody type:Primary
- Antigen name:Negative elongation factor complex member B
- Antigen symbol:NELFB
- Clonality:Monoclonal
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:25920
- Antigen synonyms:COBRA1|NELF-B
- Storage buffer:PBS (0,05% sodium azide)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to full length human COBRA1.
- Purification:Protein G purification
- Size:100 μl
- Pk:100 µg
Mouse monoclonal antibody raised against full length recombinant COBRA1.
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: NELFB
Clonality: Monoclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat