Order Entry
United Kingdom
ContactUsLinkComponent
Anti-DOK5 Goat Polyclonal Antibody
Anti-DOK5 Goat Polyclonal Antibody
Catalog # ABNOPAB6689
Supplier:  Abnova
Anti-DOK5 Goat Polyclonal Antibody
Catalog # ABNOPAB6689
Supplier:  Abnova

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    Docking Protein 5
  • Antigen symbol:
    DOK5
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Goat
  • ImmunoChemistry:
    Yes
  • Isotype:
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    55816
  • Antigen synonyms:
    IRS6|IRS-6|C20orf180
  • Storage buffer:
    Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
  • Sequence:
    CLHRTETFPAYRSEH
  • Storage temperature:
    Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    A synthetic peptide corresponding to human DOK5.
  • Purification:
    Antigen affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

Specifications

About this item

Goat polyclonal antibody raised against synthetic peptide of DOK5.

Recommended Dilutions: ELISA: 1:16000; Western Blot: 1-3 ?g/ml; Immunohistochemistry: 1-2 ?g/ml

Type: Primary
Antigen: DOK5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human