Anti-GFM2 Rabbit Polyclonal Antibody
ABNOPAB17710
: Abnova
- Antibody type:Primary
- Antigen name:G elongation factor, mitochondrial 2
- Antigen symbol:GFM2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:84340
- Antigen synonyms:mEF-G 2|RRF2mt|hEFG2|RRF2|EFG2|MRRF2|MST027|MSTP027|EF-G2mt
- Storage buffer:PBS, pH 7,4 (150mM NaCl, 0,02% sodium azide, 50% glycerol)
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to internal of human GFM2.
- Size:100 μg
- Pk:100 µg
Rabbit polyclonal antibody raised against synthetic peptide of GFM2.
Recommended Dilutions: Western Blot: 1:500-1:1000; Immunofluorescence: 1:500-1:1000; ELISA: 1:20000
Type: Primary
Antigen: GFM2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human