Anti-MELK Goat Polyclonal Antibody
Catalog # ABNOPAB6013
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Maternal Embryonic Leucine Zipper Kinase
- Antigen symbol:MELK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:9833
- Antigen synonyms:hMELK|Protein kinase PK38|KIAA0175|Maternal embryonic leucine zipper kinase|MELK|hPK38
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:KRLVEDILSSCKV
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to human MELK.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Goat polyclonal antibody raised against synthetic peptide of MELK.
Recommended Dilutions: ELISA: 1:32000; Western Blot: 0.1-0.3 ?g/ml
Type: Primary
Antigen: MELK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Human