Order Entry
United Kingdom
ContactUsLinkComponent
Anti-STOM Mouse Monoclonal Antibody [clone: 13]
Anti-STOM Mouse Monoclonal Antibody [clone: 13]
  ABNOMAB9974
 :  Abnova
Anti-STOM Mouse Monoclonal Antibody [clone: 13]
  ABNOMAB9974
 :  Abnova

 

  • Antibody type:
    Primary
  • Antigen name:
    Stomatin
  • Antigen symbol:
    STOM
  • Clonality:
    Monoclonal
  • Clone:
    13
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG2b
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Format:
    Liquid
  • Gene ID:
    2040
  • Antigen synonyms:
    STOM|EPB7|protein 7.2b|EPB72|erythrocyte membrane protein band 7.2|erythrocyte band 7 integral membrane protein|BND7
  • Storage buffer:
    Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
  • Storage temperature:
    Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
  • Immunogen:
    Recombinant protein corresponding to full length human STOM.
  • Purification:
    Affinity purification
  • Size:
    100 μg
  • Pk:
    100 µg

 

 

Mouse monoclonal antibody raised against full length recombinant STOM.

Recommended Dilutions: Western Blot: 1:1000

Type: Primary
Antigen: STOM
Clonality: Monoclonal
Clone: 13
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human