Anti-ADK Mouse Monoclonal Antibody [clone: 7]
ABNOMAB9979
: Abnova
- Antibody type:Primary
- Antigen name:Adenosine Kinase
- Antigen symbol:ADK
- Clonality:Monoclonal
- Clone:7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:132
- Antigen synonyms:Adenosine 5'-phosphotransferase|AK
- Amino acid number:138 to 362
- Storage buffer:Citrate-Tris-HCl buffer, pH 7.0 (0,02% Procl300)
- Storage temperature:Store at 4 °C. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:Recombinant protein corresponding to amino acids 138-362 of human ADK.
- Purification:Affinity purification
- Size:100 μg
- Pk:100 µg
Mouse monoclonal antibody raised against partial recombinant ADK.
Recommended Dilutions: Western Blot: 1:500
Type: Primary
Antigen: ADK
Clonality: Monoclonal
Clone: 7
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human