Anti-PSD3 Goat Polyclonal Antibody
ABNOPAB7437
: Abnova
- Antibody type:Primary
- Antigen name:Pleckstrin And Sec7 Domain Containing 3
- Antigen symbol:PSD3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Gene ID:23362
- Antigen synonyms:EFA6R|HCA67|EFA6D|NogoA
- Amino acid number:327 to 339
- Storage buffer:Tris saline, pH 7,3 (0,5% BSA, 0,02% sodium azide)
- Sequence:C-QRTASPDSKESSK
- Storage temperature:Store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to amino acids 327-339 of human PSD3.
- Purification:Antigen affinity purification
- Size:100 μg
- Pk:100 µg
Goat polyclonal antibody raised against synthetic peptide of PSD3.
Recommended Dilutions: ELISA: 1:16000
Type: Primary
Antigen: PSD3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: