Anti-TRAK2 Rabbit Polyclonal Antibody
Catalog # ABNOPAB19768
Supplier: Abnova
Specifications
- Antibody type:Primary
- Antigen name:Trafficking Protein, Kinesin Binding 2
- Antigen symbol:TRAK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Liquid
- Form:Serum
- Gene ID:66008
- Antigen synonyms:OIP98|CALS-C|AI480836|GRIF1|O-Linked N-Acetylglucosamine Transferase Interacting Protein 98|Als2cr3|AW046271|Gamma-Aminobutyric Acid(A) Receptor-Interacting Factor|Amyotrophic Lateral Sclerosis 2 Chromosomal Region Candidate Gene 3|mKIAA0549|GRIF-1
- Storage buffer:0,02% sodium azide
- Storage temperature:Store at 4 °C for three months. For long term storage store at –20 °C. Aliquot to avoid repeated freezing and thawing.
- Immunogen:A synthetic peptide corresponding to 13 amino acids near C-terminus of human TRAK2.
- Purification:Protein A purification
- Size:100 μg
- Pk:100 µg
Specifications
About this item
Rabbit polyclonal antibody raised against synthetic peptide of TRAK2.
- Recommended Dilutions: ELISA: 1:10000-80000; Western Blot: 1:200-500
Type: Primary
Antigen: TRAK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human