Anti-CKS1B Rabbit Polyclonal Antibody
ABGEAP17157C
: Abgent
- Antibody type:Primary
- Antigen name:CDC28 Protein Kinase Regulatory Subunit 1B
- Antigen symbol:CKS1B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:internal
- Gene ID:1163
- Antigen synonyms:ckshs1|CKS1|PNAS-16|PNAS-18
- Amino acid number:3 to 30
- Storage buffer:pH 7,4
- Molecular weight:9.66 kDa
- Sequence:EYRHVMLPKDIAKC
- Storage temperature:Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
- Concentration:0.5
- Shipping temperature:4 °C
- Immunogen:This CKS1B antibody is generated from rabbits immunized with a KLH conjugated synthetic peptide between 3-30 amino acids from the Central region of human CKS1B.
- Purification:Protein A & Peptide affinity purification
- Pk:400 µl
Type: Primary
Antigen: CKS1B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: internal
Host: Rabbit
Isotype:
Reactivity: Human