Order Entry
United Kingdom
ContactUsLinkComponent
Anti-CCR7 Mouse Monoclonal Antibody [clone: 617CT2.2.1]
Anti-CCR7 Mouse Monoclonal Antibody [clone: 617CT2.2.1]
Catalog # ABGEAM2119A
Supplier:  Abgent
Anti-CCR7 Mouse Monoclonal Antibody [clone: 617CT2.2.1]
Catalog # ABGEAM2119A
Supplier:  Abgent

Specifications

  • Antibody type:
    Primary
  • Antigen name:
    chemokine (C-C motif) receptor 7
  • Antigen symbol:
    CCR7
  • Clonality:
    Monoclonal
  • Clone:
    617CT2.2.1
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Mouse
  • Isotype:
    IgG1
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Epitope:
    N-terminal
  • Format:
    Ascites
  • Gene ID:
    1236
  • Antigen synonyms:
    CDw197|EBI1|BLR2|CCR-7|CD197|CMKBR7|CC-CKR-7
  • Amino acid number:
    11 to 37
  • Storage buffer:
    pH 7,4
  • Molecular weight:
    42.874 kDa
  • Sequence:
    DEVTDDYIGDNTC
  • Storage temperature:
    Maintain refrigerated at 2-8 °C for up to 6 months. For long term storage store at –20 °C in small aliquots to prevent freeze-thaw cycles
  • Shipping temperature:
    4 °C
  • Immunogen:
    This CCR7 antibody is generated from mice immunized with a KLH conjugated synthetic peptide between 11-37 amino acids from the N-terminal region of human CCR7.
  • Pk:
    0,1 mL

Specifications

About this item

Type: Primary
Antigen: CCR7
Clonality: Monoclonal
Clone: 617CT2.2.1
Conjugation: Unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG1
Reactivity: Human