Order Entry
United Kingdom
ContactUsLinkComponent
Anti-FTO Rat Monoclonal Antibody [clone: FT 342-1]
Anti-FTO Rat Monoclonal Antibody [clone: FT 342-1]
Supplier:  Cayman Chemical
Certificates
Anti-FTO Rat Monoclonal Antibody [clone: FT 342-1]
Supplier:  Cayman Chemical

Specifications

  • Catalog No:
  • CAYM10815-100
  • CAYM10815-50
  • Antigen symbol:
  • FTO
  • FTO
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • FT 342-1
  • FT 342-1
  • Reactivity:
  • Rat, Mouse
  • Rat, Mouse
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG2a kappa
  • IgG2a kappa
  • Western blot:
  • Yes
  • Yes
  • ELISA:
  • Yes
  • Yes
  • Immunogen:
  • recombinant mouse FTO
  • recombinant mouse FTO
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, pH 7,4
  • PBS, pH 7,4
  • Storage temperature:
  • –20 °C
  • –20 °C
  • Shipping temperature:
  • –20 °C
  • –20 °C

Specifications

Frequently Bought Together