Anti-Y Rabbit Polyclonal Antibody
Catalog # ROCK600-401-262
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:Phosphotyrosine
- Antigen symbol:pY
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide,
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:This Anti-PhosphoTyrosine antibody was prepared from whole rabbit serum produced by repeated immunizations with phosphotyrosine conjugated KLH.
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Anti-PhosphoTyrosine (pY) Antibody is suitable for use in ELISA, western blotting and immunoprecipitation assays. Anti-PhosphoTyrosine (pY) Antibody reacts specifically with phosphotyrosine and shows minimal reactivity by ELISA and western blotting.
Recommended Dilutions: ELISA: 1:1000-1:5000; Western Blot: 1:500; Immunochemistry: 1:50; Immunoprecipitation: 10.0 µg/ 500 µg sample;
Type: Primary
Antigen: Y
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat