Anti-ERD2 Mouse Monoclonal Antibody (Unconjugated) [clone: 2E7]
ROCK200-301-F44
- Antibody type:Primary
- Antigen symbol:ERD2
- Clonality:Monoclonal
- Clone:2E7
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2b
- Reactivity:Yeast,Drosophila,Beet,Sorghum,Mung bean,Wheat,Cotton,Barnyard grass,Saccharomyces
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-terminal
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid (sterile filtered)
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,09% (w/v) Sodium Azide, 50% (v/v) Glycerol
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:HDEL Antibody was produced in mice by repeated immunizations raised against a synthetic HDEL synthetic peptide corresponding to the C-terminus of yeast Bip.
- Purification:Protein G purified
- Size:100 µg
- Pk:100 µG
Anti-HDEL Antibody is recommended for use in WB and IF microscopy. Expect a band approximately ~78kDa corresponding to specific lysates. Specific conditions for reactivity should be optimized by the end user.
Recommended Dilutions: Immunofluorescence: 1:50-1:500; Western Blot: 1:1000-1:2000
Type: Primary
Antigen: ERD2
Clonality: Monoclonal
Clone: 2E7
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Mouse
Isotype: IgG2b
Reactivity: Drosophilia, Yeast, Saccharomyce, Barnyard Grass, Beet, Cotton, Mung Bean, Sorghum, Wheat, Yeast