Anti-ABCA4 Mouse Monoclonal Antibody (Unconjugated) [clone: 3F4]
ROCK200-301-D05
- Antibody type:Primary
- Antigen name:ATP-binding cassette, sub-family A (ABC1), member 4
- Antigen symbol:ABCA4
- Clonality:Monoclonal
- Clone:3F4
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Xenopus,Human,Bovine,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Antibody fraction
- Form:Liquid
- Gene ID:11304
- Antigen synonyms:Abcr|D430003I15Rik|Abc10|AW050280|RmP
- Storage buffer:0,01 M HEPES, 0,15 M Sodium Chloride, pH 7,50,1 mg/ml Bovine Serum Albumin - IgG and Protease free, 50% (v/v) Glycerol
- Molecular weight:220 kDa
- Storage temperature:–20 °C
- Shipping temperature:Dry Ice
- Immunogen:Anti-ABCA4 antibody was produced by repeated immunizations with partially purified bovine 220-kDa disc rim Protein.
- Purification:Protein G purified
- Size:100 µl
- Pk:100 µl
Anti-ABCA4 antibody is suitable for use in ELISA, Western Blotting and IHC. Specific conditions should be optimized by the end user. Expect a band of ~250kDa in size corresponding to ABCA4 Proteins in the appropriate cell lysate or extract.
Recommended Dilutions: ELISA: 1:10000; Western Blot: 1:1000; Immunochemistry: 1:100
Type: Primary
Antigen: ABCA4
Clonality: Monoclonal
Clone: 3F4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human, Mouse, Xenopus, Bovine