Anti-GST Mouse Polyclonal Antibody (Unconjugated) [clone: 3D4]
ROCK200-301-200
- Antibody type:Tag
- Antigen name:Glutathione-S-Transferase
- Antigen symbol:GST
- Clonality:Monoclonal
- Clone:3D4
- Conjugation:Unconjugated
- ELISA:Yes
- Gel shift:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Multiple
- Western blot:Yes
- Environmentally Preferable:
- ChIP:Yes
- Neutralisation:Yes
- Size:500 µg
- Pk:500 µG
Type: Primary
Antigen: GST
Clonality: Polyclonal
Clone: 3D4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: pan