Anti-ZBTB7B Rabbit Polyclonal Antibody
Catalog # ROCK100-401-A21
Supplier: Rockland Immunochemicals
Specifications
- Antibody type:Primary
- Antigen name:zinc finger and BTB domain containing 7B
- Antigen symbol:ZBTB7B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Internal
- Cross adsorption:No
- Format:Antiserum
- Form:Lyophilized
- Gene ID:22724
- Antigen synonyms:Thpok|c-Krox|Zfp67
- Storage buffer:0,02 M Potassium Phosphate, 0,15 M Sodium Chloride, pH 7,2, 0,01% (w/v) Sodium Azide
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:This antibody was prepared from whole rabbit serum produced by repeated immunizations with a synthetic peptide corresponding to an internal sequence of mouse cKrox Protein.
- Purification:delipidated and defibrinated antiserum
- Size:100 µl
- Pk:100 µl
Specifications
About this item
This antibody has been tested for use in ELISA, immunochemistry and western blotting. Specific conditions for reactivity should be optimized by the end user. Expect a band approximately 80 kDa in size corresponding to cKrox by western blotting.
Recommended Dilutions: ELISA: 1:2500 - 1:10000; Western Blot: 1:500 - 1:3000; Immunochemistry: 1.25-2.5 µg/ml;
Restore with deionized water (or equivalent); 100 µl
Type: Primary
Antigen: ZBTB7B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: Center
Host: Rabbit
Isotype:
Reactivity: Mouse, Human, Rat, Mouse