Anti-p21 Mouse Monoclonal Antibody [clone: WA-1 (HJ21)]
Recommended dilution: ; Immunofluorescence: 1-2?g/ml; Western Blot: 0.5-1 ?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Immunochemistry: 0.5-1 ?g/ml
Type: Primary
Antigen: p21
Clonality: Monoclonal
Clone: WA-1 (HJ21)
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Monkey, Mouse, Rat,
- Catalog No:
- USBI138579
- USBI138579-100
- Antigen symbol:
- P21
- P21
- Antibody type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- WA-1 (HJ21)
- WA-1 (HJ21)
- Reactivity:
- Monkey, Human, Rat, Mouse
- Monkey, Human, Rat, Mouse
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western blot:
- Yes
- Yes
- ELISA:
- Yes
- Yes
- ImmunoChemistry:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow cytometry:
- Yes
- Yes
- Immunogen:
- Recombinant protein corresponding to human p21.
- Recombinant protein corresponding to human p21.
- Form:
- liquid
- liquid
- Purification:
- purified by Protein G chromatography
- purified by Protein G chromatography
- Concentration:
- ~0,2 mg/ml
- ~0,2 mg/ml
- Cross adsorption:
- No
- No
- Storage buffer:
- PBS, 0,05% BSA, 0,05% sodium azide.
- PBS, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:
- –20 °C
- –20 °C
Frequently Bought Together