Anti-Cytokeratin, HMW Mouse Monoclonal Antibody [clone: AE-3]
Catalog # USBI138537-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Cytokeratin, HMW
- Clonality:Monoclonal
- Clone:AE-3
- Conjugation:Unconjugated
- ELISA:Yes
- Flow cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Rabbit,Monkey,Chicken,Human,Bovine,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, 0,05% BSA, 0,05% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,2 mg/ml
- Immunogen:Human epidermal keratin
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Recommended dilution: ;Western Blot: 0.5-1 ?g/ml; Immunofluorescence: 1-2?g/ml; Immunochemistry: 0.5-1 ?g/ml
Type: Primary
Antigen: Cytokeratin, HMW
Clonality: Monoclonal
Clone: AE-3
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Bovine, Dog, Chicken, Human, Monkey, Mouse, Rabbit, Rat,