Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Chromogranin A Mouse Monoclonal Antibody [clone: CGA414]
  USBI138532-100
Anti-Chromogranin A Mouse Monoclonal Antibody [clone: CGA414]
  USBI138532-100

 

  • Antibody type:
    Primary
  • Antigen name:
    Chromogranin A
  • Clonality:
    Monoclonal
  • Clone:
    CGA414
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Flow cytometry:
    Yes
  • Host:
    Mouse
  • ImmunoChemistry:
    Yes
  • ImmunoFluorescence:
    Yes
  • ImmunoPrecipitation:
    Yes
  • Isotype:
    IgG1 kappa
  • Reactivity:
    Human
  • Western blot:
    Yes
  • Environmentally Preferable:
  • Cross adsorption:
    No
  • Form:
    liquid
  • Storage buffer:
    PBS, 0,05% BSA, 0,05% sodium azide.
  • Storage temperature:
    –20 °C
  • Concentration:
    ~0,2 mg/ml
  • Immunogen:
    Recombinant protein fragment corresponding to human Chromogranin A.
  • Purification:
    purified by Protein G chromatography
  • Size:
    100 µg
  • Pk:
    100 µG

 

 

Recommended dilution: ;Western Blot: 0.5-1 ?g/ml; Immunofluorescence: 1-2?g/ml; Immunoprecipitation: 1-2?g/500 ?g; Immunochemistry: 0.5-1 ?g/ml

Type: Primary
Antigen: Chromogranin A
Clonality: Monoclonal
Clone: CGA414
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,