Order Entry
United Kingdom
ContactUsLinkComponent
 
Anti-Ficolin B Rat Monoclonal Antibody [clone: IA4]
 
undefined
Anti-Ficolin B Rat Monoclonal Antibody [clone: IA4]

 

  • Catalog No:
  • USBIF4499-60A-25
  • USBIF4499-60A-100
  • Antigen symbol:
  • FCNB
  • FCNB
  • Antigen name:
  • Ficolin B
  • Ficolin B
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • IA4
  • IA4
  • Reactivity:
  • Mouse
  • Mouse
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG2b
  • IgG2b
  • Western blot:
  • Yes
  • Yes
  • Immunogen:
  • Mouse ficolin B (E. coli expressed)
  • Mouse ficolin B (E. coli expressed)
  • Form:
  • liquid
  • liquid
  • Purification:
  • purified by immunoaffinity chromatography
  • purified by immunoaffinity chromatography
  • Concentration:
  • ~0,5 mg/ml
  • ~0,5 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, pH 7,2, 0,09% sodium azide.
  • PBS, pH 7,2, 0,09% sodium azide.
  • Storage temperature:
  • –20 °C
  • –20 °C

 

 

Frequently Bought Together