Anti-Cyclin A2 Mouse Monoclonal Antibody [clone: E23.1]
USBIC8452-06C-25
- Antibody type:Primary
- Antigen name:Cyclin A2
- Clonality:Monoclonal
- Clone:E23.1
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human,Bovine,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:–20 °C
- Concentration:~0,5 mg/ml
- Immunogen:Cyclin A fusion protein
- Purification:purified by immunoaffinity chromatography
- Size:25 μg
- Pk:25 µG
Recommended dilution: Western Blot: 0.5-1 ?g/ml
Type: Primary
Antigen: Cyclin A2
Clonality: Monoclonal
Clone: E23.1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Bovine, Human, Mouse,