Anti-Calcitonin Sheep Polyclonal Antibody
USBIC0115-08F-100
- Antibody type:Primary
- Antigen name:Calcitonin
- Antigen symbol:CALCA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Sheep
- Isotype:IgG
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Format:Serum
- Form:lyophilized powder
- Sequence:CGNLSTCMLGTYTQDFNKFHTFPQTAIGVGAP
- Storage temperature:–20 °C
- Immunogen:Synthetic peptide corresponding to CGNLSTCMLGTYTQDFNKFHTFPQTAIGVGAP from human Calcitonin, comjugated to BSA
- Tested applications:Radio Immunoassay
- Pk:100 µl
Recommended dilution: Radio Immunoassay: 1:80000
Type: Primary
Antigen: Calcitonin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Sheep
Isotype: IgG
Reactivity: Human,