Anti-BD2 Mouse Monoclonal Antibody [clone: L12-4C-C2]
Catalog # USBIB0901-01B-10
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen symbol:BD2
- Clonality:Monoclonal
- Clone:L12-4C-C2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Environmentally Preferable:
- Cross adsorption:No
- Form:lyophilized powder
- Amino acid number:4 to 41
- Storage buffer:50mM Tris, pH 7,4.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Synthetic human β-Defensin 2 (aa 4-41) (DPVTCLKSGAICHPVFCPRRYKQIGTCGLPGTKCCKKP)
- Purification:purified by Protein G chromatography
- Size:10 μg
- Pk:10 µG
Specifications
About this item
Type: Primary
Antigen: BD2
Clonality: Monoclonal
Clone: L12-4C-C2
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human,