Anti-BD1 Rabbit Polyclonal Antibody
USBIB0899-03L-100
- Antibody type:Primary
- Antigen symbol:BD1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Format:Serum
- Form:lyophilized powder
- Amino acid number:18 to 26
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Synthetic human β-Defensin 1 (aa 18-26) (PIFTKIQGT)
- Pk:100 µl
Type: Primary
Antigen: BD1
Clonality: Polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human,