Anti-PDCD4 Mouse Monoclonal Antibody [clone: 8j9]
USBIP3120-67-100
- Antibody type:Primary
- Antigen name:Programmed cell death 4
- Antigen symbol:PDCD4
- Clonality:Monoclonal
- Clone:8j9
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Recombinant human PDCD4 (1-469aa) purified from E. coli
- Purification:purified by Protein G chromatography
- Size:100 μl
- Pk:100 µl
Recommended dilution: Western Blot: 1:2000-1:3000
Type: Primary
Antigen: PDCD4
Clonality: Monoclonal
Clone: 8j9
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,