Anti-Hexokinase, Type III Mouse Monoclonal Antibody [clone: 8G7]
Catalog # USBIH2040-30C-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Hexokinase 3
- Antigen symbol:HK3
- Clonality:Monoclonal
- Clone:8G7
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG1 kappa
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS, pH 7,4, 0,1% sodium azide.
- Storage temperature:–20 °C
- Concentration:~1 mg/ml
- Immunogen:Recombinant human Hexokinase 3 (1-923aa) purified from E. coli
- Purification:purified by Protein G chromatography
- Size:100 μl
- Pk:100 µl
Specifications
About this item
Recommended dilution: Western Blot: 1:1000-1:2000
Type: Primary
Antigen: Hexokinase, Type III
Clonality: Monoclonal
Clone: 8G7
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,