Order Entry
United Kingdom
ContactUsLinkComponent
Anti-Glucose Regulated Protein 94 Rat Monoclonal Antibody [clone: 7L6]
Anti-Glucose Regulated Protein 94 Rat Monoclonal Antibody [clone: 7L6]
Supplier:  US Biological
Certificates
Anti-Glucose Regulated Protein 94 Rat Monoclonal Antibody [clone: 7L6]
Supplier:  US Biological

Specifications

  • Catalog No:
  • USBIG3057-40B-200
  • USBIG3057-40B-50
  • Antigen symbol:
  • GRP94
  • GRP94
  • Antigen name:
  • Glucose Regulated Protein 94
  • Glucose Regulated Protein 94
  • Antibody type:
  • Primary
  • Primary
  • Clonality:
  • Monoclonal
  • Monoclonal
  • Conjugation:
  • Unconjugated
  • Unconjugated
  • Clone:
  • 7L6
  • 7L6
  • Reactivity:
  • Dog, Rat, Mouse, Hamster, Pig, Rabbit, Horse, Monkey, Xenopus, Sheep, Chicken, Human, Bovine, Guinea pig
  • Dog, Rat, Mouse, Hamster, Pig, Rabbit, Horse, Monkey, Xenopus, Sheep, Chicken, Human, Bovine, Guinea pig
  • Host:
  • Rat
  • Rat
  • Isotype:
  • IgG2a
  • IgG2a
  • Western blot:
  • Yes
  • Yes
  • ImmunoPrecipitation:
  • Yes
  • Yes
  • Flow cytometry:
  • Yes
  • Yes
  • Immunogen:
  • Purified Grp94 isolated from chicken oviducts
  • Purified Grp94 isolated from chicken oviducts
  • Form:
  • liquid
  • liquid
  • Purification:
  • purified by Protein G chromatography
  • purified by Protein G chromatography
  • Concentration:
  • ~1 mg/ml
  • ~1 mg/ml
  • Cross adsorption:
  • No
  • No
  • Storage buffer:
  • PBS, pH 7,2, 0,09% sodium azide and 50% glycerol.
  • PBS, pH 7,2, 0,09% sodium azide and 50% glycerol.
  • Storage temperature:
  • –20 °C
  • –20 °C

Specifications

Related Information

Frequently Bought Together