Anti-Phosphoserine Mouse Monoclonal Antibody [clone: 6D662]
Catalog # USBIP4076-20-100
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Phosphoserine
- Antigen symbol:pS
- Clonality:Monoclonal
- Clone:6D662
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgM
- Reactivity:Human,Dog,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Form:liquid
- Storage buffer:PBS containing 0,09% sodium azide, PEG, andsucrose.
- Storage temperature:–20 °C
- Concentration:~0,1 mg/ml
- Immunogen:Phosphoserine containing peptides.
- Purification:purified by thiophilic adsorption and size exclusion chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Type: Primary
Antigen: Phosphoserine
Clonality: Monoclonal
Clone: 6D662
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgM
Reactivity: Dog, Human, Mouse, Rat,