Anti-14-3-3 GRF Rabbit Polyclonal Antibody
Catalog # AGRIAS122119
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:14-3-3 GRF
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:E. pulcherrima
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- UniProtKB:Q9C5W6
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:conserved surface-exposed peptide conjugated to KLH.This sequence is repeated in all 12 “canonical” members of the Arabidopsis thaliana GRF/14-3-3 family. GRF1 GF14 chi
- Size:50 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: 14-3-3 GRF
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: E. pulcherrima