Anti-SPS Rabbit Polyclonal Antibody
Catalog # AGRIAS06185
Supplier: Agrisera
Specifications
- Antibody type:Primary
- Antigen symbol:SPS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:S. lycopersicum,Zea mays
- Western blot:Yes
- Environmentally Preferable:
- Cross adsorption:No
- Storage temperature:–20 °C
- Shipping temperature:Ambient
- Immunogen:Synthetic peptide derived from Zea mays SPS protein sequence (P31927).
- Size:100 µl
- Pk:50 µl
Specifications
About this item
Type: Primary
Antigen: SPS
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: S. lycopersicum, Zea mays