Anti-TMX1 Mouse Monoclonal Antibody [clone: 1F7]
ORIGTA507001
: OriGene
- Antibody type:Primary
- Antigen symbol:TMX1
- Clonality:Monoclonal
- Clone:1F7
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:No
- Form:Liquid
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:31.6 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human TMX1(NP_110382) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μl
- Pk:100 µl
Recommended Dilutions: Western Blot: 1:1000
Type: Primary
Antigen: TMX1
Clonality: Monoclonal
Clone: 1F7
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG1
Reactivity: Human