Anti-ARL2BP Mouse Monoclonal Antibody [clone: 4E8]
Catalog # ORIGCF800364
Supplier: OriGene
Specifications
- Antibody type:Primary
- Antigen name:ADP-ribosylation factor-like 2 binding Protein
- Antigen symbol:ARL2BP
- Clonality:Monoclonal
- Clone:4E8
- Conjugation:Unconjugated
- Host:Mouse
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:Full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (PH 7.3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:18.6 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human ARL2BP (NP_036238) produced in E.coli.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Specifications
About this item
Recommended Dilutions: Western Blot: 1:2000
Type: Primary
Antigen: ARL2BP
Clonality: Monoclonal
Clone: 4E8
Conjugation: Unconjugated
Epitope: Full length
Host: Mouse
Isotype: IgG2a
Reactivity: Human