Anti-NEUROG1 Mouse Monoclonal Antibody [clone: 19A11]
ORIGCF500309
: OriGene
- Antibody type:Primary
- Antigen name:Neurogenin 1
- Antigen symbol:NEUROG1
- Clonality:Monoclonal
- Clone:19A11
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Epitope:full length
- Cross adsorption:No
- Form:Lyophilized
- Storage buffer:PBS (pH 7,3) containing 1% BSA, 50% glycerol and 0,02% sodium azide.
- Molecular weight:25.7 kDa
- Storage temperature:–20 °C
- Shipping temperature:–20 °C
- Immunogen:Full length human recombinant protein of human NEUROG1 (NP_006152) produced in HEK293T cell.
- Purification:purified by affinity chromatography
- Size:100 μg
- Pk:100 µG
Recommended Dilutions: Western Blot: 1:2000; Immunochemistry: 1:50; Immunoprecipitation: 2-4?g/mg
Type: Primary
Antigen: NEUROG1
Clonality: Monoclonal
Clone: 19A11
Conjugation: Unconjugated
Epitope: full length
Host: Mouse
Isotype: IgG1
Reactivity: Human