Anti-RTN3 Rabbit Antibody
Catalog # BSBTPA2256
Supplier: Boster Bio
Specifications
- Antibody type:Primary
- Antigen name:Reticulon 3
- Antigen symbol:RTN3
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western blot:Yes
- Environmentally Preferable:
- Epitope:C-Terminal
- Cross adsorption:No
- Form:Lyophilised
- Antigen synonyms:RTN3|RTN3-A1|NSPL2|HAP|ASYIP|NSPLII
- Amino acid number:1017 - 1032
- Sequence:EKIQAKLPGIAKKKAE
- Storage temperature:–20 °C
- Immunogen:A synthetic peptide corresponding to a sequence at the C-terminus of human RTN3 (1017-1032aa EKIQAKLPGIAKKKAE), identical to the related mouse and rat sequences.
- Purification:purified by immunogen affinity purification
- Size:100 µg
- Pk:100 µG
Specifications
About this item
Rabbit IgG polyclonal antibody for Reticulon-3(RTN3) detection. Tested with WB, IHC-P in Human, Mouse, Rat.
Reconstitute with distilled water
Add 0,2 ml of distilled water will yield a concentration of 500 µg/ml.
Type: Primary
Antigen: RTN3
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope: C-Terminal
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat