Anti-Transforming Growth Factor, beta Binding Protein 2, latent Mouse Monoclonal Antibody [clone: 9G2]
Catalog # USBIT8252-01
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:Transforming Growth Factor, beta Binding Protein 2, latent
- Clonality:Monoclonal
- Clone:9G2
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoPrecipitation:Yes
- Isotype:IgG
- Reactivity:Bovine
- Western blot:Yes
- Environmentally Preferable:
- Epitope:N-terminal
- Format:ascites
- Form:lyophilized powder
- Storage buffer:0,01% thimerosal.
- Storage temperature:Cool
- Shipping temperature:Ice
- Size:200 µl
- Pk:200 µl
Specifications
About this item
Recommended dilution: ELISA: 1:1000 -1:2000; Western Blot: 1:200 -1:500; Immunoprecipitation: 1:100 -1:200; Immunochemistry: 1:50 -1:100
Type: Primary
Antigen: Transforming Growth Factor, beta Binding Protein 2, latent
Clonality: monoclonal
Clone: 9G2
Conjugation: unconjugated
Epitope: N-terminal
Host: Mouse
Isotype: IgG
Reactivity: Bovine