Anti-Toxoplasma gondii Mouse Monoclonal Antibody [clone: 11H64]
USBIT8076-10J
- Antibody type:Primary
- Antigen symbol:Toxoplasma gondii
- Clonality:Monoclonal
- Clone:11H64
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG1
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,1% sodium azide.
- Storage temperature:Cool
- Shipping temperature:Ice
- Purification:purified by Protein A chromatography
- Size:1 mg
- Pk:1 mg
Type: Primary
Antigen: Toxoplasma gondii
Clonality: monoclonal
Clone: 11H64
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: