Anti-THEMIS Goat Polyclonal Antibody
USBIT3773-20A
- Antibody type:Primary
- Antigen name:Thymocyte selection associated
- Antigen symbol:THEMIS
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Goat
- Isotype:
- Reactivity:Mouse
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:Tris saline, 0,02% sodium azide, pH 7,3, 0,5% BSA.
- Sequence:C-EERTINLPKSLKSH
- Storage temperature:Cool
- Concentration:~0,5 mg/ml
- Shipping temperature:Ice
- Purification:purified by immunoaffinity chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:64000; Western Blot: 1-3?g/ml
Type: Primary
Antigen: THEMIS (thymocyte selection associated)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Goat
Isotype:
Reactivity: Mouse