Anti-STABILIN-1-INTERACTING CHITINASE-LIKE PROTEIN, Rat Monoclonal Antibody [clone: 10F432]
Catalog # USBIS7950-56A
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:STABILIN-1-INTERACTING CHITINASE-LIKE PROTEIN,
- Clonality:Monoclonal
- Clone:10F432
- Conjugation:Unconjugated
- Flow cytometry:Yes
- Host:Rat
- Isotype:IgG2a
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2, 0,09% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:200 µg
- Pk:200 µG
Specifications
About this item
Type: Primary
Antigen: STABILIN-1-INTERACTING CHITINASE-LIKE PROTEIN,
Clonality: Monoclonal
Clone: 10F432
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Human