Anti-SCO Cytochrome Oxidase Deficient Homolog 2 Rabbit Polyclonal Antibody
Catalog # USBIS0509-20
Supplier: US Biological
Specifications
- Antibody type:Primary
- Antigen name:SCO Cytochrome Oxidase Deficient Homolog 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- Host:Rabbit
- Isotype:
- Reactivity:Human
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, 0,02% sodium azide.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Size:50 µg
- Pk:50 µG
Specifications
About this item
Recommended dilution: Immunochemistry: 2.5 ?g/ml
Type: Primary
Antigen: SCO Cytochrome Oxidase Deficient Homolog 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human