Anti-Rotavirus VP6 Mouse Monoclonal Antibody [clone: 2A27A]
USBIR3004-05A
- Antibody type:Primary
- Antigen name:Rotavirus VP6
- Clonality:Monoclonal
- Clone:2A27A
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Mouse
- Isotype:IgG2b
- Reactivity:
- Western blot:Yes
- Environmentally Preferable:
- Form:liquid
- Storage buffer:PBS, pH 7,2.
- Storage temperature:Cool
- Concentration:~1 mg/ml
- Shipping temperature:Ice
- Purification:purified by Protein G chromatography
- Size:100 µg
- Pk:100 µG
Recommended dilution: ELISA: 1:50000; Western Blot: 1:5000
Type: Primary
Antigen: Rotavirus VP6
Clonality: monoclonal
Clone: 2A27A
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: